missing translation for 'onlineSavingsMsg'
Learn More

CD161 Mouse anti-Human, FITC, Clone: DX12, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 556080

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 15808638

  • 340.00€ / Each


Estimated Shipment: 08-10-2021
Add to basket

Description

Description

The DX12 monoclonal antibody specifically binds to human CD161 which is also known as Natural killer cell surface protein P1A (NKR-P1A). CD161 is encoded by the KLRB1 (Killer cell lectin-like receptor subfamily B member 1) gene. CD161 is a member of the C-type lectin superfamily and is also referred to as C-type lectin domain family 5 member B (CLEC5B). CD161 is a 80 kDa disulfide-linked homodimer, type II membrane glycoprotein. CD161 is expressed mostly on NK cell populations and on subsets of CD4+ and CD8+ αβ T cells, NKT cells and γδ T cells. Reports indicate that CD161 is expressed preferentially on CD45RO+ T cells, however, it can be found on a subset of thymocytes and fetal liver T cells. Its function has not been fully elucidated, but reports indicate that NKR-P1A may serve as a specific receptor for some NK cell targets. The DX12 antibody can reportedly inhibit spontaneous cytotoxicity mediated by certain NK cell clones.

Specifications

Specifications

CD161
DX12
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Mouse
IgG1, κ
RUO
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Flow Cytometry
FITC
Affinity Purified
Human NKR-P1A Transfected Mouse Fibroblasts
100 Tests
Immunology
Primary
Human
Videos
SDS
Documents
Certificates

For Research Use Only.