missing translation for 'onlineSavingsMsg'
Learn More

ACE/Angiotensin Converting Enzyme FITC Monoclonal Antibody, Clone: i1A8, Invitrogen™

ACE/Angiotensin Converting Enzyme Antibody, FITC conjugate (i1A8), MA5-16722, from Invitrogen™. Species Reactivity: Human, Non-human primate; Applications: Flow Cytometry.

Brand:  Invitrogen MA516722

Additional Details : Weight : 0.01000kg

Product Code. 15265979

  • 362.00€ / 100µg


Estimated Shipment date
from Supplier 23-03-2020
Add to basket

Description

Description

This gene encodes an enzyme involved in catalyzing the conversion of angiotensin I into a physiologically active peptide angiotensin II. Angiotensin II is a potent vasopressor and aldosterone-stimulating peptide that controls blood pressure and fluid-electrolyte balance. This enzyme plays a key role in the renin-angiotensin system. Many studies have associated the presence or absence of a 287 bp Alu repeat element in this gene with the levels of circulating enzyme or cardiovascular pathophysiologies. Two most abundant alternatively spliced variants of this gene encode two isozymes - the somatic form and the testicular form that are equally active. Multiple additional alternatively spliced variants have been identified but their full length nature has not been determined.
Host Species: Mouse
Cross Reactivity: Human, Non-human primate
Isotype: IgG1
Clone: i1A8
Immunogen: Human lung CD143 (Angiotensin converting enzyme)
Formulation: 100 ug purified antibody (0.1mg/mL) in PBS with 1% BSA and 0.09% sodium azide
Synonyms: ACE1, CD143, DCP, DCP1, MGC26566, MVCD3
Uniprot Accession: P12821
Entrez GeneID: 1636
Gene Symbol: ACE
For FACS analysis, use 10ul of the suggested working dilution to label 1x10^6 cells in 100ul.
Specifications

Specifications

ACE
Monoclonal
FITC
P12821
ACE1, CD143, DCP, DCP1, MGC26566, MVCD3
Murine
IgG1
Store at 4°C or Store at -20°C if preferred, store in dark
Primary
1636
Flow Cytometry
i1A8
100μg purified antibody (0.1mg/mL) in PBS with 1% BSA and 0.09% sodium azide
Purified
ACE
Human lung CD143 (Angiotensin converting enzyme)
100μg
Primary
Monoclonal
Human, Non-human Primate
Videos
Documents