missing translation for 'onlineSavingsMsg'
Learn More

CD14 Mouse anti-Human, BUV737, Clone: M5E2, BD

Mouse Monoclonal Antibody

251.94€ - 520.20€

Specifications

Antigen CD14
Clone M5E2
Applications Flow Cytometry
Classification Monoclonal
Conjugate Brilliant Ultraviolet 737
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15823649
View Documents Promotion Details
BD Biosciences
564445
25 Tests This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
N/A View alternative products
Save Up to 
»
15813649
View Documents Promotion Details
BD Biosciences
564444
100 Tests This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
N/A View alternative products
Save Up to 
»
Description

Description

The M5E2 monoclonal antibody specifically binds to CD14, a 53–55 kDa glycosylphosphatidylinositol (GPI)-anchored single chain glycoprotein expressed at high levels on monocytes. Additionally, the anti-CD14 antibody reacts with interfollicular macrophages, reticular dendritic cells, and some Langerhans cells. CD14 has been identified as a high affinity cell-surface receptor for complexes of lipopolysaccharide (LPS) and serum LPS-binding protein, LPB.

The antibody was conjugated to BD Horizon™ BUV737 which is part of the BD Horizon Brilliant™ Ultraviolet family of dyes. This dye is a tandem fluorochrome of BD Horizon BUV395 with an Ex Max of 348-nm and an acceptor dye with an Em Max at 737-nm. BD Horizon Brilliant BUV737 can be excited by the ultraviolet laser (355 nm) and detected with a 740/35 filter. Due to the excitation of the acceptor dye by other laser lines, there may be significant spillover into channels detecting Alexa Fluor™ 700-like dyes (e.g., 712/20-nm filter).

Specifications

Specifications

CD14
Flow Cytometry
Brilliant Ultraviolet 737
RUO
Human CD14 Protein
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
M5E2
Monoclonal
Mouse
Baboon, Canine, Cynomolgus, Human, Rhesus Monkey
IgG2a κ
Affinity Purified
LPS receptor; LPS-R; Myeloid cell-specific leucine-rich glycoprotein
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.