missing translation for 'onlineSavingsMsg'
Learn More

CD14 Mouse anti-Human, Alexa Fluor 488, Clone: M5E2, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 561706

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15811259

  • 258.00€ / Each


Estimated Shipment: 07-10-2021
Add to basket

Description

Description

The M5E2 monoclonal antibody specifically binds to CD14, a 53–55 kDa glycosylphosphatidylinositol (GPI)-anchored single chain glycoprotein expressed at high levels on monocytes. Additionally, the anti-CD14 antibody reacts with interfollicular macrophages, reticular dendritic cells, and some Langerhans cells. CD14 has been identified as a high affinity cell-surface receptor for complexes of lipopolysaccharide (LPS) and serum LPS-binding protein, LPB.

This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Specifications

Specifications

CD14
M5E2
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Mouse
IgG2a, κ
RUO
Primary
Baboon, Canine, Cynomolgus, Human, Rhesus Monkey
Flow Cytometry
Alexa Fluor 488
Affinity Purified
Human CD14 Protein
25 Tests
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Videos
SDS
Documents
Certificates

For Research Use Only.