Learn More
CCR6 Mouse anti-Human, Clone: 53103.111, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA120156
Additional Details : Weight : 0.01000kg
Description
MA1-20156 detects CCR6 in human samples. MA1-20156 has been successfully used in FACS and immunohistochemistry (frozen) procedures. The MA1-20156 immunogen is human CCR6 transfected NSO mouse myeloma cells. Endotoxin level is less than 10 ng per mg antibody as determined by the LAL method.
Chemokines have been sub-divided into families on the basis of the relative position of their cysteine residues. The a- and b- families, with four cysteine residues, are the largest and best characterized. CCR-6 (previously referred to as the GPR-CY4, CKRL3, or STRL22 orphan receptor) binds MIP-3". It is a CC chemokine receptor that is highly expressed in human dendritic cells derived from CD34+ cord blood precursors.2-4 In addition to dendritic cells, CCR-6 mRNA has also been detected in lymphocytes (CD4 and CD8 T cells and B cells), but not in natural killer cells, monocytes and granulocytes.Specifications
CCR6 | |
Monoclonal | |
0.5 mg/mL | |
CCR6 | |
BN-1, CKR-L3, CKR6, CKRL3, CMKBR6, DCR2, DRY-6, GPR-CY4, GPR29, GPRCY4, STRL22 | |
Mouse | |
Protein G | |
RUO | |
1235 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Flow Cytometry, Immunohistochemistry (Frozen) | |
53103.111 | |
Unconjugated | |
P51684 | |
CCR6 | |
Human CCR6 transfected NSO mouse myeloma cells. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |