missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carboxypeptidase A5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA539080
Additional Details : Weight : 0.00900kg
Description
Carboxypeptidases have functions ranging from digestion of food to selective biosynthesis of neuroendocrine peptides. Members of the A/B subfamily of carboxypeptidases, such as CPA5, contain an approximately 90-amino acid pro region that assists in the folding of the active carboxypeptidase domain. Cleavage of the pro region activates the enzyme.Specifications
Carboxypeptidase A5 | |
Polyclonal | |
Unconjugated | |
CPA5 | |
Carboxypeptidase A5 , CPA5 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
74649, 93979 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q8WXQ8, Q8R4H4 | |
CPA5 | |
A synthetic peptide derived from the internal region of human CPA5 | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |