Learn More
C20orf194 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA540433
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: FVNFFGDKTD FHPLMDQFMN DYVEEANREI EKYNQELEQQ EYHDLFELKP Sequence homology: Cow: 93%; Dog: 93%; Horse: 93%; Human: 100%; Mouse: 86%; Pig: 93%; Rabbit: 93%; Rat: 86%; Yeast: 83%
C20orf194 Polyclonal antibody specifically detects C20orf194 in Human samples. It is validated for Western Blot.Specifications
C20orf194 | |
Polyclonal | |
Unconjugated | |
C20orf194 | |
C20orf194; chromosome 20 open reading frame 194; RGD1565616; Uncharacterized protein C20orf194; uncharacterized protein C20orf194 homolog | |
Rabbit | |
Protein A | |
RUO | |
25943 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q5TEA3 | |
C20orf194 | |
Synthetic peptide directed towards the C-terminal of human C20ORF194 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |