Learn More
BFSP2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA545632
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: CQQVGEAVLE NARLMLQTET IQAGADDFKE RYENEQPFRK AAEEEINSLY Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 86%
BFSP2 Polyclonal antibody specifically detects BFSP2 in Human samples. It is validated for Western Blot.Specifications
BFSP2 | |
Polyclonal | |
Unconjugated | |
BFSP2 | |
49 kDa cytoskeletal protein; AI448593; beaded filament protein CP49; beaded filament structural protein 2; beaded filament structural protein 2, phakinin; bfps2, Cytoskeletal protein, 49-kD; BFSP2; CP 47; CP47; CP49; CTRCT12; DKEYP-73D8.2; hypothetical protein LOC494090; lens fiber cell beaded filament protein CP 47; Lens fiber cell beaded filament protein CP 49; lens intermediate filament-like light; LIFL-L; MGC142078; MGC142080; phakinin; PHAKOSIN; zgc:103750 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
8419 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q13515 | |
BFSP2 | |
Synthetic peptide directed towards the middle region of human BFSP2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |