missing translation for 'onlineSavingsMsg'
Learn More

Bcl-X Mouse, Unlabeled, Clone: 4, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 610210

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15825339

  • 421.00€ / 1 set


Estimated Shipment: 08-10-2021
Add to basket

Description

Description

Bcl-x is related to the Bcl-2 protein and can function independently of Bcl-2 in regulating apoptosis (programmed cell death). The bcl-x transcript is found in a number of tissues, with the highest levels in the lymphoid and central nervous systems. Two distinct cDNA species, bcl-xL and bcl-xS, have been observed and appear to arise from alternate 5′ splice sites located within the first coding exon of the bcl-x gene. Bcl-xL is composed of 233 amino acids and is similar in size and structure to Bcl-2. Bcl-xS lacks 63 amino acids corresponding to the region of Bcl-xL with the highest degree of homology to Bcl-2. Like Bcl-2, Bcl-xL has been reported to inhibit cell death upon growth factor withdrawal in an IL-3-dependent cell line. However, Bcl-xS inhibits the ability of Bcl-2 to enhance cell survival in the absence of growth factors.

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

Specifications

Specifications

Bcl-X
4
Unconjugated
Affinity Purified
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
150μg
Cell Biology
Primary
Chicken, Murine, Rat
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Mouse
IgG2b
RUO
Store undiluted at -20°C.
Monoclonal
Videos
SDS
Documents
Certificates

For Research Use Only.