missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATF5 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA517988
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with mouse and rat based on sequence homology.
ATF5 Polyclonal antibody specifically detects ATF5 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
ATF5 | |
Polyclonal | |
Unconjugated | |
Atf5 | |
activating transcription factor 5; Activating transcription factor 5-alpha/beta; activating transcription factor 7; activating transcription factor X; AFTA; ATF5; Atf7; Atfx; BZIP protein ATF7; cAMP-dependent transcription factor ATF-5; Cyclic AMP-dependent transcription factor ATF-5; FLJ34666; HMFN0395; NAP1; NRIF3-associated protein; ODA-10; Transcription factor ATFx; transcription factor-like protein ODA-10 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
107503, 22809, 282840 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
O70191, Q6P788, Q9Y2D1 | |
Atf5 | |
Synthetic peptide sequence (EVYKARSQRTRSC) corresponding to the C-terminus amino acids of ATF5 | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |