missing translation for 'onlineSavingsMsg'
Learn More

Aquaporin 12 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA536426

Additional Details : Weight : 0.00910kg

Product Code. 15587661

  • 471.00€ / 100µL
Estimated Shipment: 17-05-2024
to see stock.
Add to basket

Description

Description

This antibody detects endogenous protein at a molecular weight of 31 kDa. Purity is >95% by SDS-PAGE.

Aquaporin 12 Polyclonal antibody specifically detects Aquaporin 12 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications

Specifications

Aquaporin 12
Polyclonal
Unconjugated
AQP12A
AQP12; AQP-12; AQP12A; AQP12B; AQP-12B; AQPX2; aquaporin 12; aquaporin 12A; aquaporin 12B; aquaporin X2; aquaporin-12; aquaporin-12A; Aquaporin-12B; INSSA3; insulin synthesis associated 3; RGD1562727
Rabbit
Antigen affinity chromatography
RUO
208760, 367316, 375318, 653437
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
1 mg/mL
PBS with 0.05% sodium azide; pH 7.2
A6NM10, Q8CHJ2, Q8IXF9
Aqp12, AQP12A, AQP12B
Synthetic peptide corresponding to amino acids 230-296 of human AQP12
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers