missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Androgen Receptor Mouse anti-Human, Clone: H7507, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen 415900
Additional Details : Weight : 0.50000kg
Description
The androgen receptor (AR) is a member of the steroid-hormone receptor superfamily of nuclear receptors. The receptor is more than 90 kDa and has three major functional domains: the N-terminal domain, DNA-binding domain, and the androgen-binding domain. The androgen receptor is a ligand-activated transcription factor that binds active testosterone (T) and dihydrotestosterone (DHT). Upon binding the hormone ligand, the receptor dissociates from accessory proteins, translocates into the nucleus, dimerizes, and then stimulates transcription of androgen responsive genes. The AR signaling pathway plays a key role in development and function of male reproductive organs, including the prostate and epididymis. AR also plays a role in nonreproductive organs, such as muscle, hair follicles, and brain. Androgen Receptor is a phosphoprotein, and also regulates mitogen-activated protein kinase (MAP kinase). The inhibition of the MEK1/2 pathway correlates directly with a change in phosphorylation state of the androgen receptor. Abnormalities in the AR signaling pathway have been linked to a number of diseases, including prostate cancer, Kennedy′s disease, and male infertility. Mutations in this gene are associated with complete androgen insensitivity (CAIS).Specifications
| Androgen Receptor | |
| Monoclonal | |
| 1 mg/mL | |
| physiological saline with 0.1% sodium azide | |
| P10275 | |
| DHTR, NR3C4, Dihydrotestosterone receptor, Nuclear receptor subfamily 3 group C member 4, AR | |
| Mouse | |
| IgG2a | |
| 100 μg | |
| -20°C | |
| Primary | |
| 367 |
| ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot | |
| H7507 | |
| Unconjugated | |
| AR | |
| Liquid | |
| AR | |
| Produced in BALB/c mouse ascites after inoculation with hybridoma of mouse myeloma cells (NS-1) and spleen cells derived from a BALB/c mouse immunized with Baculovirus-expressed recombit human AR (2-53 aa). | |
| Ammonium sulfate precipitation | |
| RUO | |
| Antibody | |
| Monoclonal | |
| Human |