missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AMHR2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA513902
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with mouse and rat based on sequence homology.
AMHR2 Polyclonal antibody specifically detects AMHR2 in Human samples. It is validated for Flow Cytometry, Western Blot.Specifications
AMHR2 | |
Polyclonal | |
Unconjugated | |
AMHR2 | |
AMH type II receptor; AMHR; Amhr2; Anti-Muellerian hormone type II receptor; anti-Muellerian hormone type-2 receptor; anti-Mullerian hormone receptor type 11 SV1; anti-Mullerian hormone receptor type 2; anti-Mullerian hormone receptor type II; anti-Mullerian hormone receptor, type II; anti-Mullerian hormone type 2 receptor; anti-Mullerian hormone type 2 receptor delta 2; anti-Mullerian hormone type 2 receptor delta 9/10; C14; MIS type II receptor; Misiir; MISR2; MISRII; MRII; Muellerian inhibiting substance type II receptor; Mullerian inhibiting substance type II receptor | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
269 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Western Blot | |
2 mg/mL | |
PBS with 0.09% sodium azide | |
Q16671 | |
AMHR2 | |
KLH conjugated synthetic peptide between 374-402 amino acids from the C-terminal region of human AMHR2 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |