Learn More
Catenin alpha-1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA528345
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: 293T, A431, mouse brain, rat brain. Predicted reactivity: Mouse (100%), Rat (100%), Zebrafish (93%), Xenopus laevis (96%), Rabbit (100%), Rhesus Monkey (99%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Catenin alpha-1 Polyclonal antibody specifically detects Catenin alpha-1 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.Specifications
Catenin alpha-1 | |
Polyclonal | |
Unconjugated | |
CTNNA1 | |
[a]E-catenin; 102 kDa cadherin-associated protein; 2010010M04Rik; AA517462; AI988031; alpha E catenin; alpha E-catenin; alpha(E)-catenin; alpha-E-catenin; cadherin associated protein; Cadherin-associated protein; CAP102; catenin (cadherin associated protein), alpha 1; catenin (cadherin-associated protein), alpha; catenin (cadherin-associated protein), alpha 1; catenin (cadherin-associated protein), alpha 1, 102kDa; catenin alpha 1; catenin alpha 1 S homeolog; catenin alpha-1; catenin, alpha 1; Catna1; cell-adhesion protein alphaE catenin; ctnna; ctnna1; ctnna1.S; MDPT2; Renal carcinoma antigen NY-REN-13; XELAEV_18020018mg | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
12385, 1495, 307505 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
1 mg/mL | |
PBS with 20% glycerol and 0.01% thimerosal, 0.025% ProClin 300; pH 7 | |
P26231, P35221 | |
CTNNA1 | |
Recombinant fragment corresponding to a region within amino acids 511 and 906 of Human alpha Catenin | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |