Learn More
AKR7A3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA528482
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: 293T, HeLaS3, HepG2, Molt-4, Raji, Mammary gland tumor tissues. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
AKR7A3 Polyclonal antibody specifically detects AKR7A3 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
AKR7A3 | |
Polyclonal | |
Unconjugated | |
AKR7A3 | |
Afar; AFAR2; AFB1 aldehyde reductase 2; AFB1-AR; AFB1-AR 2; aflatoxin aldehyde reductase; aflatoxin B1 aldehyde reductase 2; aflatoxin B1 aldehyde reductase member 1; Aflatoxin B1 aldehyde reductase member 3; Akr7a1; AKR7A3; aldo-keto reductase family 7 member A3; aldo-keto reductase family 7, member A3 (aflatoxin aldehyde reductase); rAFAR1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
22977 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
O95154 | |
AKR7A3 | |
Recombinant fragment corresponding to a region within amino acids 139 and 331 of Human AKR7A3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |