Learn More
Anisonitrile, 99%, Thermo Scientific Chemicals
Catalog Number p-60607
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
874-90-8
C8H7NO
133.15
MFCD00001818
XDJAAZYHCCRJOK-UHFFFAOYSA-N
anisonitrile, p-anisonitrile, 4-cyanoanisole, benzonitrile, 4-methoxy, p-methoxybenzonitrile, 4-anisonitrile, p-anisylnitrile, p-cyanoanisole, p-methoxyphenyl cyanide, 4-methoxy benzonitrile
70129
4-methoxybenzonitrile
COC1=CC=C(C=C1)C#N
874-90-8
C8H7NO
133.15
MFCD00001818
XDJAAZYHCCRJOK-UHFFFAOYSA-N
anisonitrile, p-anisonitrile, 4-cyanoanisole, benzonitrile, 4-methoxy, p-methoxybenzonitrile, 4-anisonitrile, p-anisylnitrile, p-cyanoanisole, p-methoxyphenyl cyanide, 4-methoxy benzonitrile
70129
4-methoxybenzonitrile
COC1=CC=C(C=C1)C#N
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point 57°C to 60°C
Color White to Beige
Boiling Point 256°C to 257°C (765.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula CH3OC6H4CN
Quantity 25 g
Beilstein 10, 168
Solubility Information Solubility in water: soluble
Formula Weight 133.15
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Anisonitrile, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Anisonitrile
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.