missing translation for 'onlineSavingsMsg'
Learn More

Honeywell Fluka™ Ammonium thiocyanate concentrate, For 1L standard solution, 0.1 M NH4SCN (0.1N), Honeywell Fluka™

for 1L standard solution, 0.1 M NH4SCN (0.1N)

Brand:  Honeywell Fluka™ 38020-6X1EA

Additional Details : CAS : 1762-95-4 Weight : 0.51000kg

Quantity 6 x 1 ea.

 View more versions of this product

Product Code. 15625190

  • 178.92€ / Pack of 6


EU Stock 10
Add to basket

Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

1762-95-4
76.117
SOIFLUNRINLCBN-UHFFFAOYSA-N
15666
C(#N)[S-].[NH4+]
CH4N2S
MFCD00011428
ammonium thiocyanate, ammoniumrhodanid, thiocyanic acid, ammonium salt, ammonium isothiocyanate, ammonium rhodanate, weedazol tl, trans-aid, ammonium sulfocyanate, ammonium sulfocyanide, rhodanine, ammonium salt
azanium;thiocyanate

Specifications

Ammonium thiocyanate concentrate
7732-18-5
<20.0000%
NH4SCN
UN3082
ammonium thiocyanate, ammoniumrhodanid, thiocyanic acid, ammonium salt, ammonium isothiocyanate, ammonium rhodanate, weedazol tl, trans-aid, ammonium sulfocyanate, ammonium sulfocyanide, rhodanine, ammonium salt
C(#N)[S-].[NH4+]
76.117
76.12
Ampoule
6 x 1 ea.
For 1L standard solution, 0.1 M NH4SCN (0.1N)
≥10.0000%
CH4N2S
MFCD00011428
3595135
SOIFLUNRINLCBN-UHFFFAOYSA-N
azanium;thiocyanate
15666
Concentrate
100°C
Videos
SDS Documents