missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Sulfate, ACS, ≥99%, Ultrapure, Thermo Scientific Chemicals

Catalog Number 15815258
Click to view available options
Quantity:
1 kg
5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy
7783-20-2
H8N2O4S
132.13
MFCD00003391
BFNBIHQBYMNNAN-UHFFFAOYSA-N
ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate
6097028
CHEBI:62946
diazanium;sulfate
N.N.OS(O)(=O)=O
This item is not returnable. View return policy
7783-20-2
H8N2O4S
132.13
MFCD00003391
BFNBIHQBYMNNAN-UHFFFAOYSA-N
ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate
6097028
CHEBI:62946
diazanium;sulfate
N.N.OS(O)(=O)=O

Used in purification of enzymes and other proteins by precipitation

Used in the purification of enzymes and other proteins by precipitation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point ∼280°C (decomposition)
Physical Form Powder
Quantity 1 kg
Linear Formula (NH4)2SO4
Formula Weight 132.14
Percent Purity ≥99%
Grade ACS
Packaging Plastic bottle
Specific Gravity 1.769
Density 1.769
Chemical Name or Material Ammonium Sulfate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'