missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Sulfamate, BioXtra, ≥98%, Honeywell Fluka™

Catalog Number p-7120194
Click to view available options
Quantity:
25 g
100 g
Packaging:
Poly Bottle
This item is not returnable. View return policy
7773-06-0
H6N2O3S
114.119
MFCD00011429
GEHMBYLTCISYNY-UHFFFAOYSA-N
ammonium sulfamate, ammonium sulphamate, ammate, ikurin, ammonium amidosulfonate, feliderm k, ammate x, sulfamic acid, monoammonium salt, ammonium sulfamidate, ammonium sulphamidate
24482
CHEBI:81950
azanium;sulfamate
[NH4+].NS(=O)(=O)[O-]
This item is not returnable. View return policy
7773-06-0
H6N2O3S
114.119
MFCD00011429
GEHMBYLTCISYNY-UHFFFAOYSA-N
ammonium sulfamate, ammonium sulphamate, ammate, ikurin, ammonium amidosulfonate, feliderm k, ammate x, sulfamic acid, monoammonium salt, ammonium sulfamidate, ammonium sulphamidate
24482
CHEBI:81950
azanium;sulfamate
[NH4+].NS(=O)(=O)[O-]

BioXtra,≥98.0%

  • Stringent testing protocols may include trace metal analysis and specialized suitability testing, such as hybridoma or mouse embryo
  • Includes application-specific formulations, such as tablets and prepared solutions.
TRUSTED_SUSTAINABILITY
Melting Point 131°C to 135°C
Color White
pH 4.5 to 6.0 (at 25°C)
Quantity 25 g
Linear Formula NH4SO3NH2
UN Number UN3077
Formula Weight 114.12g/mol
Percent Purity ≥98%
Packaging Poly Bottle
Appearance Crystalline
Chemical Name or Material Ammonium sulfamate
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'