missing translation for 'onlineSavingsMsg'
Learn More

Ammonium sodium phosphate tetrahydrate, 99%, for analysis, Thermo Scientific Chemicals

Catalog Number p-3756603
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
Packaging:
Plastic bottle
7783-13-3
H5NNaO44H2O
209.07
MFCD00066970
IQTQISLCLWJRPM-UHFFFAOYSA-M
unii-k78u8wlv4h, ammonium sodium phosphate tetrahydrate, k78u8wlv4h, phosphorsalz, acmc-20ajp4, ammonium sodium phosphate tetrahydrate mi, sodium monoammonium monohydrogen phosphate tetrahydrate, phosphoric acid, monoammonium monosodium salt, tetrahydrate, salt of phosphorus microcosmic salt, sodium ammonium phosphate tetrahydrate
56924540
azanium;sodium;hydrogen phosphate;tetrahydrate
[NH4+].O.O.O.O.OP(=O)([O-])[O-].[Na+]
7783-13-3
H5NNaO44H2O
209.07
MFCD00066970
IQTQISLCLWJRPM-UHFFFAOYSA-M
unii-k78u8wlv4h, ammonium sodium phosphate tetrahydrate, k78u8wlv4h, phosphorsalz, acmc-20ajp4, ammonium sodium phosphate tetrahydrate mi, sodium monoammonium monohydrogen phosphate tetrahydrate, phosphoric acid, monoammonium monosodium salt, tetrahydrate, salt of phosphorus microcosmic salt, sodium ammonium phosphate tetrahydrate
56924540
azanium;sodium;hydrogen phosphate;tetrahydrate
[NH4+].O.O.O.O.OP(=O)([O-])[O-].[Na+]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For analysis
TRUSTED_SUSTAINABILITY
Melting Point 80°C
Color Colorless to White
Physical Form Crystalline Powder and/or Crystals
Quantity 250 g
Assay Percent Range 99%
Linear Formula Na(NH4)HPO4·4H2O
Merck Index 12,587
Formula Weight 209.07
Percent Purity 99%
Grade Analytical
Packaging Plastic bottle
Specific Gravity 1.54
Density 1.5400g/mL
Chemical Name or Material Ammonium sodium phosphate tetrahydrate, For analysis
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'