missing translation for 'onlineSavingsMsg'
Learn More

Ammonium iron(III) sulfate dodecahydrate, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., 99.0-102.0%, crystalline, slightly violet, Honeywell Fluka™

Catalog Number p-7071991
Click to view available options
Quantity:
500 g
2.5 kg
50 kg
7783-83-7
FeH28NO20S2
482.176
LCPUDZUWZDSKMX-UHFFFAOYSA-K
ammonium ferric sulfate, ferric ammonium sulfate dodecahydrate, iron ammonium sulfate dodecahydrate, sulfuric acid, ammonium iron 3+ salt 2:1:1 , dodecahydrate, ferric ammonium sulfate, acs, azanium iron 3+ disulfate dodecahydrate, ferric iron ammonium dodecahydrate disulfate, azanium iron +3 cation disulfate dodecahydrate, ammonium iron iii sulfate dodecahydrate, acs 100g
197097
azanium;iron(3+);disulfate;dodecahydrate
[NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Fe+3]
7783-83-7
FeH28NO20S2
482.176
LCPUDZUWZDSKMX-UHFFFAOYSA-K
ammonium ferric sulfate, ferric ammonium sulfate dodecahydrate, iron ammonium sulfate dodecahydrate, sulfuric acid, ammonium iron 3+ salt 2:1:1 , dodecahydrate, ferric ammonium sulfate, acs, azanium iron 3+ disulfate dodecahydrate, ferric iron ammonium dodecahydrate disulfate, azanium iron +3 cation disulfate dodecahydrate, ammonium iron iii sulfate dodecahydrate, acs 100g
197097
azanium;iron(3+);disulfate;dodecahydrate
[NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Fe+3]

Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., 99.0-102.0%, crystalline, slightly violet

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Color Violet
Physical Form Crystalline
Quantity 500 g
Linear Formula NH4Fe(SO4)2 · 12H2O
MDL Number MFCD00150004
UN Number NONH for all modes of transport
Solubility Information In accordance with water, 10g/dL
Formula Weight 482.19g/mol
Percent Purity 99 to 102%
Grade ACS/puriss. p.a.
Chemical Name or Material Ammonium Iron(Iii) Sulfate Dodecahydrate, Reag. Ph. Eur. reag. ISO
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'