missing translation for 'onlineSavingsMsg'
Learn More

Ammonium iron(III) sulfate dodecahydrate, ACS, 98.5-102.0%, Thermo Scientific Chemicals

Catalog Number p-4407224
Click to view available options
Quantity:
500 g
2.5 kg
7783-83-7
FeH28NO20S2
482.176
MFCD00150004
LCPUDZUWZDSKMX-UHFFFAOYSA-K
ammonium ferric sulfate, ferric ammonium sulfate dodecahydrate, iron ammonium sulfate dodecahydrate, sulfuric acid, ammonium iron 3+ salt 2:1:1 , dodecahydrate, ferric ammonium sulfate, acs, azanium iron 3+ disulfate dodecahydrate, ferric iron ammonium dodecahydrate disulfate, azanium iron +3 cation disulfate dodecahydrate, ammonium iron iii sulfate dodecahydrate, acs 100g
197097
azanium;iron(3+);disulfate;dodecahydrate
[NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Fe+3]
7783-83-7
FeH28NO20S2
482.176
MFCD00150004
LCPUDZUWZDSKMX-UHFFFAOYSA-K
ammonium ferric sulfate, ferric ammonium sulfate dodecahydrate, iron ammonium sulfate dodecahydrate, sulfuric acid, ammonium iron 3+ salt 2:1:1 , dodecahydrate, ferric ammonium sulfate, acs, azanium iron 3+ disulfate dodecahydrate, ferric iron ammonium dodecahydrate disulfate, azanium iron +3 cation disulfate dodecahydrate, ammonium iron iii sulfate dodecahydrate, acs 100g
197097
azanium;iron(3+);disulfate;dodecahydrate
[NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Fe+3]

Ammonium iron(III) sulfate dodecahydrate is used in dyeing and printing textiles, as analytical lab reagent. It is often employed as an analytical standard, and has been used in a variety of other applications from nanomaterials to general redox reactions. It plays an important role for the inactivation of mycobacteriophage D29 bacteria which is vivale for viable mycobacterium smegmatis and m.tuberculosis. The crystalline ammonium ferrous sulfate has shown useful function as the high absorbed dose and low-temperature dosimeter.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 39°C to 41°C
Color Purple
Physical Form Broken Crystals
Quantity 500 g
Assay Percent Range 98.5-102.0%
Solubility Information Freely soluble in water and hydrochloric acid. Insoluble in alcohol.
Formula Weight 482.18 (266.00 Anhydrous)
Grade ACS
Odor Odorless
Appearance Pale purple broken crystals
Density 1.71 g/cm3
Chemical Name or Material Ammonium iron(III) sulfate dodecahydrate, For analysis ACS
Show More Show Less
compliance-icons
Product Identifier
  • Ammonium iron(III) sulfate dodecahydrate
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P264-Wash thoroughly after handling.
  • P362-Take off contaminated clothing.
  • P321-Specific treatment (see on this label).
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'