missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Hydroxide, 28.0 to 30.0%, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
Glass Bottle
Specifications
Quantity | 500 mL |
IUPAC Name | azanium;hydroxide |
Formula Weight | 35.05 |
Percent Purity | 28.0 to 30.0% |
Grade | BAKER ANALYZED™ ACS Reagent |
Packaging | Glass Bottle |
Specific Gravity | 1L = 0.90kg |
Chemical Name or Material | Ammonium Hydroxide |
Safety and Handling
Signal Word
- Danger
- Acute toxicity Category 4
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1
- Specific target organ toxicity Category 3
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
- H400-Very toxic to aquatic life.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'