missing translation for 'onlineSavingsMsg'
Learn More

Ammonia Solution, 7N in Methanol, Honeywell Fluka™

Catalog Number p-7120162
Click to view available options
Quantity:
100 mL
1 L
Packaging:
Glass Bottle
This item is not returnable. View return policy
7664-41-7
H3N
17.03
MFCD00011418
QGZKDVFQNNGYKY-UHFFFAOYSA-N
ammonia, ammonia gas, spirit of hartshorn, nitro-sil, ammoniakgas, ammonia anhydrous, anhydrous ammonia, ammonia, anhydrous, ammoniak, am-fol
222
CHEBI:16134
N
This item is not returnable. View return policy
7664-41-7
H3N
17.03
MFCD00011418
QGZKDVFQNNGYKY-UHFFFAOYSA-N
ammonia, ammonia gas, spirit of hartshorn, nitro-sil, ammoniakgas, ammonia anhydrous, anhydrous ammonia, ammonia, anhydrous, ammoniak, am-fol
222
CHEBI:16134
N

7N in methanol

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Color Colorless
Physical Form Liquid
Quantity 100 mL
Linear Formula NH3
UN Number UN3286
Flash Point 14°C (57.2°F)
IUPAC Name ammonia
Formula Weight 17.03g/mol
Concentration or Composition (by Analyte or Components) 7N in methanol
Packaging Glass Bottle
Density 0.779g/mL at 25°C
Chemical Name or Material Ammonia solution
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'