missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonia Solution, 0.5M in Dioxane, Honeywell™
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
Glass Bottle
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Color | Colorless |
Linear Formula | NH3 |
Concentration | 0.5M in dioxane |
UN Number | UN1165 |
Density | 1.023g/mL at 25°C |
Quantity | 100 mL |
Flash Point | 11°C (51.8°F) |
Formula Weight | 17.03g/mol |
Packaging | Glass Bottle |
Vapor Pressure | 4.16psi (20°C), 11.73psi (55°C) |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'