Learn More
VACUUBRAND™ Aluminum Reducing Pieces With Small KF Flanges
Brand:  VACUUBRAND™ 20669043
45.70 EUR valid until 2020-12-31
Use promo code "14720" to get your promotional price.
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 10029440
  • 49.85€ / Each
Estimated Shipment
23-11-2020
Add to basket
Specifications
Specifications
40/16 mm
Aluminum
40 mm
Solenoid-operated valves VV and VV C
1 Each
Reducing Pieces