Learn More
SCAT Air Valve for SafetyCap, Fire Resistant, UNF 1/4 in. Thread
Fire-resistant air filter suitable for a waste container safety cap port.
Brand:  SCAT 112010
Additional Details : Weight : 0.25000kg
Product Code. 10271985
  • 40.23€ / Each
please call +351214253350
Add to basket
Description
Description
  • Fire-resistant
  • Should be changed every 6 months
  • Filters up to 150 mL/min.
  • UNF 1/4 in. thread
Specifications
Specifications
150 mL/min. filtration
Air filtration
Waste container safety cap
Air Filter