Learn More
Thermo Scientific Chemicals Agarose LM, low melt, for recovery of samples after separation
Description
Used for size-based separation of nucleic acids in agarose gel electrophoresis applications. Ideal for analysis and recovery of DNA and RNA for routine applications. Ideal for a variety of applications such as in-gel manipulation, tissue and cell culture, and viral plaque assays. Can also be used in protein electrophoresis applications such as Ouchterlony (antigen-antibody interaction assay) and radial immunodiffusion (RID) (antigen quantitation assay)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Molecular Formula | C24H38O19 |
Product Type | LM Agarose |
InChI Key | MJQHZNBUODTQTK-WKGBVCLCSA-N |
SMILES | OC[C@H]1O[C@@H](O[C@@H]2[C@@H]3CO[C@H]2[C@H](O)[C@H](O[C@H]2[C@@H](O)[C@@H](CO)O[C@@H](O[C@@H]4[C@@H]5CO[C@H]4[C@H](O)[C@H](O)O5)[C@@H]2O)O3)[C@H](O)[C@@H](O)[C@H]1O |
IUPAC Name | (2S,3R,4S,5R,6R)-2-{[(1S,3S,4S,5S,8R)-3-{[(2S,3R,4S,5S,6R)-2-{[(1S,3R,4S,5S,8R)-3,4-dihydroxy-2,6-dioxabicyclo[3.2.1]octan-8-yl]oxy}-3,5-dihydroxy-6-(hydroxymethyl)oxan-4-yl]oxy}-4-hydroxy-2,6-dioxabicyclo[3.2.1]octan-8-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol |
Molecular Weight (g/mol) | 630.55 |
Quantity | 5 g |
Sensitivity | Hygroscopic; Moisture sensitive |
RUO – Research Use Only