missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Adenosine 5'-triphosphate disodium salt hydrate Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
10 g
25 g
100 g
500 g
Packaging:
Plastic bottle
Specifications
Physical Form | Crystalline Powder |
Quantity | 25 g |
Formula Weight | 551.14 (Anhydrous) |
Packaging | Plastic bottle |
Chemical Name or Material | Adenosine-5 '-Triphosphate Disodium Salt Trihydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'