Learn More
Socorex™ Acura™ Manual TwiXS Micropipette Twin Pack
Superior, extra sharp micropipettes that are short, smooth and lightweight. Socorex™ Acura™ Manual TwiXS Micropipette Twin Pack provides you with a select pair of two ergonomically designed micropipettes that are optimally suited for research laboratories.
605.00€ - 625.00€
Specifications
Autoclavable | Autoclavable |
---|---|
Model | Manual XS |
Number of Channels | 1 |
Product Line | Acura |
Type | Micropipette Twin Pack |
Product Code | Brand | Accuracy | Precision | Volume (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Accuracy | Precision | Volume (Metric) | Price | Quantity & Availability | |||
11787966
|
Socorex™
826.TWX.E |
<±6.0% - <±2.0% / <±2.5% - <±1.0% | <5.0% - <1.5% / <1.7% - <0.5% | 0.1 to 2, 2 to 20 μL |
625.00€
1 set |
Estimated Shipment: 01-07-2024
Log in to see stock. |
|||
11707976
|
Socorex™
826.TWX.K |
<±1.5% - <±1.0% / <±1.5% - <±0.6% | <1.0% - <0.4% / <0.6% - <0.2% | 5 to 50, 20 to 200 μL |
605.00€
1 set |
Estimated Shipment: 08-08-2024
Log in to see stock. |
|||
11898423
|
Socorex™
826.TWX.H |
<±2.5% - <±1.0% / <±1.5% - <±0.6% | <1.7% - <0.5% / <0.6% - <0.2% | 2 to 20, 20 to 200 μL |
605.00€
1 set |
Estimated Shipment: 08-08-2024
Log in to see stock. |
|||
11808433
|
Socorex™
826.TWX.Q |
<±2.5% - <±1.0% / <±1.5% - <±0.5% | <0.6% - <0.2% / <0.5% - <0.2% | 20 to 200, 100 to 1000 μL |
605.00€
1 set |
Estimated Shipment: 17-05-2024
Log in to see stock. |
|||
11717976
|
Socorex™
826.TWX.N |
<±1.5% - <±0.8% / <±1.5% - <±0.5% | <0.6% - <0.2% / <0.5% - <0.2% | 10 to 100, 100 to 1000 μL |
605.00€
1 set |
Estimated Shipment: 08-08-2024
Log in to see stock. |
|||
Description
- TwiXS Pack:
- Two Acura™ Manual XS Micropipettes.
- Free shelf pipette holder.
- Qualitips™ samples.
- QC certificates.
- Operating instructions.
- Optimized shape/size ratio for ease of use when working with micro tubes.
- Reduced weight - lighter than leading brands.
- Unique, Socorex™ patented lip seal reduces activation forces and provides for ultra-soft pipetting.
- Adjustable tip ejector.
- Autoclavable fully assembled.
- Unsurpassed metrologic performance.
Specifications
Autoclavable | |
1 | |
Micropipette Twin Pack | |
2/Pk. |
Manual XS | |
Acura | |
Twin pack | |
36 Months |