Learn More
Memmert™ Active Microprocessor Control
Memmert™ Active Microprocessor Control ensures more rapid reaching of set humidity and very short recovery times while avoiding condensate formation.
Brand:  Memmert™ ICO105K7
Additional Details : Weight : 2.80000kg
 View more versions of this product
Product Code. 15622214
  • 1045.00€ / Each
please call +351214253350
Add to basket
Description
Description
  • Humidity supply with distilled water (from an external tank) by a self-priming pump
  • Integral bacteria block by generating hotsteam
  • Dehumidifying via sterile filter
Specifications
Specifications
40 - 97 % rh
Humidity Supply
Digital indication and auto-diagnostic system
ICO105
Humidifying and dehumidifying
Videos