Learn More
Asecos™ Activated Carbon Filter
Aids in ventilation in safety storage cabinets.
779.10€ - 821.50€
Product Code | Brand | Dimensions (D x W x H) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (D x W x H) | Price | Quantity & Availability | |||||
15664844
|
Asecos™
32952 |
270 x 270 x 230 mm |
779.10€
Each |
Estimated Shipment: 16-08-2024
Log in to see stock. |
|||||
15941732
|
Asecos™
32953 |
270 x 270 x 165 mm |
821.50€
Each |
Estimated Shipment: 16-08-2024
Log in to see stock. |
|||||
Description
- Activated carbon
- Available in different sizes
- Accessory item
Specifications
Filter | |
CX, SLX models |