Learn More
Abnova™ ACSS3/CEN12p FISH Probe
Labeled FISH probes for identification of gene amplification using Fluoresecent In Situ Hybridization Technique.
Brand: Abnova™ FG0171.100uL
Additional Details : Weight : 0.02000kg
Specifications
Store at 4°C in the dark. | |
Tube | |
79611 | |
100 μL |
DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe | |
FISH-Ce | |
DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe | |
Liquid |