missing translation for 'onlineSavingsMsg'
Learn More

Acetic acid, potassium salt, 99+%, for biochemistry, Thermo Scientific Chemicals

Catalog Number 11914911
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy
127-08-2
C2H3KO2
98.14
SCVFZCLFOSHCOH-UHFFFAOYSA-M
potassium acetate, acetic acid, potassium salt, diuretic salt, potassium ethanoate, koac, octan draselny czech, potassiumacetate, acetic acid potassium salt, acok, fema no. 2920
517044
CHEBI:32029
potassium;acetate
CC(=O)[O-].[K+]
This item is not returnable. View return policy
127-08-2
C2H3KO2
98.14
SCVFZCLFOSHCOH-UHFFFAOYSA-M
potassium acetate, acetic acid, potassium salt, diuretic salt, potassium ethanoate, koac, octan draselny czech, potassiumacetate, acetic acid potassium salt, acok, fema no. 2920
517044
CHEBI:32029
potassium;acetate
CC(=O)[O-].[K+]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer
TRUSTED_SUSTAINABILITY
Melting Point 292.0°C
Flash Point >250°C
Assay Percent Range 99+%
Packaging Plastic bottle
Linear Formula CH3CO2K
Quantity 2.5 kg
Merck Index 15,7726
Solubility Information (5 M in water) Clear colorless solution
Formula Weight 98.14
Percent Purity 99+%
Grade Biochemistry
Chemical Name or Material Acetic acid, potassium salt
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'