missing translation for 'onlineSavingsMsg'
Learn More

BINDER™ Left Access Port (50 to 125 mm)

For Use With: BINDER units ED 23, ED 400

Brand:  BINDER™ 8012-0542

Additional Details : Weight : 0.50000kg

Product Code. 15621786

  • 589.00€ / Each
Estimated Shipment: 12-07-2024
to see stock.
Add to basket

Description

Description

  • For use on left
  • Includes silicone plug
  • For use with a wide range of BINDER™ incubators and chambers
Specifications

Specifications

Access port with silicone plug - 50 mm, left
Access Port
50 mm
BINDER Chambers ED 23, ED 400
Silicone plug
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers