missing translation for 'onlineSavingsMsg'
Learn More

Uvex™ 9770 Pheos E-S-WR Unventilated Safety Helmets

Lightweight helmets are suitable for use with ambient temperatures down to -30°C, and resistant to hot molten lava splash. Helmets feature a short brim for unrestricted upward vision, and a suspension harness with wheel ratchet for variable width adjustment.

Brand:  Uvex™ 9770431

Additional Details : Weight : 0.40200kg

 View more versions of this product

Product Code. 17209111

  • 19.10€ / Each
Estimated Shipment: 24-05-2024
to see stock.
Add to basket

Description

Description

  • Fully enclosed helmet shell is suitable for use by electricians
  • Fulfills all safety requirements for construction and industrial work
  • Suspension harness with wheel ratchet allows for continuous width adjustment
  • Shortened brim ensures unrestricted upward vision
  • Side Euroslot adapters (30 mm) for attaching earmuffs and visor systems
  • Six-point textile band attachment for optimal fit and comfort
  • Easily withstands pressure from the side
  • Can be used in temperatures as low as -30°C
  • Resistant to molten metal splash
  • Includes a printing area for personalization
Specifications

Specifications

EN 397:2012, EN 50365:2002
Low Voltage Systems
HDPE
Electrically Insulating Helmet
Short Brim
Not Ventilated
Green
Adapter System for Uvex™ Visors, Helmet Capsules, and Helmet Lamps
9770 Pheos E-S-WR
52 to 61 cm
Harness with Wheel Ratchet
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers