Learn More
Eppendorf™ 96-Well Flat Bottom Sterile Polypropylene Microplates
292.00€ - 790.00€
Specifications
Format | Standard |
---|---|
Array | 8 x 12 |
Color | White, Clear Bottom |
Material | Polypropylene |
No. of Wells | 96 |
Product Code | Brand | Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Price | Quantity & Availability | |||||
10658204
|
Eppendorf™
0030606108 |
White, Clear Bottom |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
10310604
|
Eppendorf™
0030602102 |
White, Clear Bottom |
292.00€
Pack of 80 |
Estimated Shipment: 23-05-2024
Log in to see stock. |
|||||
Specifications
Standard | |
White, Clear Bottom | |
96 | |
Flat Bottom (F-well) | |
Clear |
8 x 12 | |
Polypropylene | |
Sterile | |
White |