Learn More
Uvex 95284 Low Tune-Up Insole For Uvex 1 Sport - Width 11
Tune-Up antistatic insole for reducing fatigue and supporting arch
Brand:  Uvex 9528450
Additional Details : Weight : 0.16600kg
 View more versions of this product
Product Code. 17208071
  • 21.80€ / 1 pair
Estimated Shipment: 21-05-2024
to see stock.
Add to basket
Description
Description
Uvex Tune-Up Comfortable Low Insole 95284
  • Antistatic insole concept for use in safety footwear
  • Designed to reduce fatigue by increasing wearer comfort and enhancing ergonomics by optimum support of arch of feet
  • Good cushioning
  • Relief of pressure marks
  • Non-irritating, comfortable surface structure
  • Breathable and absorbs moisture
  • Antibacterial
  • Antistatic
  • Useable for ESD-footwear
  • Uvex 1 Sport
  • Article No. 95284
  • Standard EN ISO 20345:2011
  • Sizes: 35 to 52
Specifications
Specifications
Black, Blue
Microvelour
No
Standard width
No
Uvex 1 Sport
50
Unisex