missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ 900 mm ReVo™ Class II, Type A2 Microbiological Safety Cabinet

Product Code. 17840754 Shop All Labconco Corporation Products
Change view
Click to view available options
Ports:
Schuko Plug
UK Plug
Includes:
Base Stand
Base Stand Not Included
Unit Size:
1 unit
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Ports Includes Unit Size
17840754 Schuko Plug Base Stand 1 unit
17810754 Schuko Plug Base Stand Not Included 1 unit
17840744 UK Plug Base Stand Not Included 1 unit
17830754 UK Plug Base Stand 1 unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 17840754 Supplier Labconco™ Supplier No. 3R2310131
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

ReVo™ Class II, Type A2 Microbiological Safety Cabinets provide personnel, product and environmental protection from biohazardous agents and hazardous particulates.

  • Provide personnel, product and environmental protection from biohazardous agents and hazardous particulates.
  • Other appropriate applications include work with antineoplastic drugs, genetic material, and additional substances that generate hazardous airborne particulates..
  • All models are EN 12469 compliant with additional certification by TÜV Nord.
  • These biological safety cabinets are designed to discharge HEPA-filtered exhaust air directly into the laboratory, or into an exhaust system through the optional Canopy Connection.
  • When canopy-connected, the biosafety cabinets may be used for applications involving minute quantities of volatile toxic chemicals and tracer amounts of radionuclides as an adjunct to microbiological research.
  • During operation, room air is drawn into the inlet grill.
  • The air in the plenum beneath the work surface is a mixture of unfiltered room air and air that has just passed through the work area.
  • This contaminated air is drawn by the blower through the back plenum of the cabinet, where approximately 70% of the air is recirculated through the supply HEPA filter and back over the work area.
  • The balance of the contaminated air is discharged to the environment after passing through the exhaust HEPA filter.

Specifications

Certifications/Compliance EN 12469 Certified by TÜV
Chamber Material Stainless Steel (304)
Product Type Class II Biosafety Cabinet
Noise Level 43 dBA
Outlets 2
Power Consumption 85 W
Sash Opening (Metric) 200 mm
Exhaust Air Filter HEPA Exhaust Filter
Filter Type H14 HEPA
Includes Base Stand
Ports Schuko Plug
Sash Opening (English) 7.87 in.
Dimensions (L x W x H) Exterior 797 x 1004 x 1350 mm
Shipping Weight (Metric) 177.0 kg
Voltage 230 V
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.