Learn More
BINDER™ Temperature Calibration Certificate with 9 Measurement Points
For Use With: BINDER units series BD
Brand:  BINDER™ 8012-0915
Additional Details : Weight : 0.01000kg
Product Code. 15621976
  • 822.00€ / Each
Estimated Shipment date
from Supplier 23-07-2020
Add to basket
Description
Description
  • Measurement at specified temperature
  • Including certificate
  • Various points of measurement
Specifications
Specifications
Temperature Calibration Certificate
Calibration certificate, temperature - temperature measurement incl. certificate, 9 measuring points at specified temperature
BINDER units series BD
Temperature measurement