Learn More
Binder™ Calibration Certificate for Temperature
Binder™ Calibration Certificate for Temperature features a variety of measuring points at specified temperatures for a range of Binder™ incubators and chambers.
Brand:  Binder™ 8012-0915
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15621976
  • 798.00€ / Each
please call +351214253350
Add to basket
Description
Description
  • Measurement at specified temperature
  • Including certificate
  • Various points of measurement
Specifications
Specifications
9 measurement points
For use with BD 56, BD 115, BD 260 models
Temperature measurement
Temperature Calibration Certificate