missing translation for 'onlineSavingsMsg'
Learn More

8-Benzyl-3-(trifluoromethylsulfonyloxy)-8-azabicyclo[3.2.1]oct-3-ene, 95%, Thermo Scientific™

Catalog Number p-4228373
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
949902-02-7
C15H16F3NO3S
347.36
GBUXWXWPNDHMRG-UHFFFAOYSA-N
8-benzyl-8-aza-bicyclo 3.2.1 oct-2-en-3-yl trifluoromethanesulfonate, 8-benzyl-8-azabicyclo 3.2.1 oct-2-en-3-yl trifluoromethanesulfonate, 8-benzyl-8-azabicyclo 3.2.1 oct-3-en-3-yl trifluoromethanesulfonate, trifluoro-methanesulfonic acid 8-benzyl-8-aza-bicyclo 3.2.1 oct-2-en-3-yl ester, trifluoro-methanesulfonic acid 8-benzyl-8-azabicyclo 3.2.1 oct-2-en-3-yl ester, trifluoromethanesulfonic acid 8-benzyl-8-aza-bicyclo 3.2.1 oct-2-en-3-yl ester
49835910
(8-benzyl-8-azabicyclo[3.2.1]oct-3-en-3-yl) trifluoromethanesulfonate
C1CC2C=C(CC1N2CC3=CC=CC=C3)OS(=O)(=O)C(F)(F)F
This item is not returnable. View return policy
949902-02-7
C15H16F3NO3S
347.36
GBUXWXWPNDHMRG-UHFFFAOYSA-N
8-benzyl-8-aza-bicyclo 3.2.1 oct-2-en-3-yl trifluoromethanesulfonate, 8-benzyl-8-azabicyclo 3.2.1 oct-2-en-3-yl trifluoromethanesulfonate, 8-benzyl-8-azabicyclo 3.2.1 oct-3-en-3-yl trifluoromethanesulfonate, trifluoro-methanesulfonic acid 8-benzyl-8-aza-bicyclo 3.2.1 oct-2-en-3-yl ester, trifluoro-methanesulfonic acid 8-benzyl-8-azabicyclo 3.2.1 oct-2-en-3-yl ester, trifluoromethanesulfonic acid 8-benzyl-8-aza-bicyclo 3.2.1 oct-2-en-3-yl ester
49835910
(8-benzyl-8-azabicyclo[3.2.1]oct-3-en-3-yl) trifluoromethanesulfonate
C1CC2C=C(CC1N2CC3=CC=CC=C3)OS(=O)(=O)C(F)(F)F
CAS Min % 94.0
CAS Max % 100.0
Color Orange
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 347.36
Percent Purity 95%
Physical Form Viscous Liquid, May Contain Suspension
Chemical Name or Material 8-Benzyl-3-(trifluoromethylsulfonyloxy)-8-azabicyclo[3.2.1]oct-3-ene
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 8-Benzyl-3-(trifluoromethylsulfonyloxy)-8-azabicyclo¢3.2.1!oct-3-ene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'