Learn More
6-Heptynoic acid, 95%, Thermo Scientific Chemicals
Catalog Number 15505246
Click to view available options
Quantity:
1 g
5 g
25 g
30964-00-2
C7H10O2
126.16
MFCD00239430
OFCPMJGTZUVUSM-UHFFFAOYSA-N
6-heptynoic acid, 6-heptynoicacid, 6-heptynoic alcohol, ksc222a4l, ofcpmjgtzuvusm-uhfffaoysa
4377950
hept-6-ynoic acid
OC(=O)CCCCC#C
30964-00-2
C7H10O2
126.16
MFCD00239430
OFCPMJGTZUVUSM-UHFFFAOYSA-N
6-heptynoic acid, 6-heptynoicacid, 6-heptynoic alcohol, ksc222a4l, ofcpmjgtzuvusm-uhfffaoysa
4377950
hept-6-ynoic acid
OC(=O)CCCCC#C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Quantity 25 g
Formula Weight 126.16
Percent Purity 95%
Chemical Name or Material 6-Heptynoic acid
compliance-icons
Product Identifier
  • 6-Heptynoic acid, 95%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.