Learn More
Tocris Bioscience™ 5J 4
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
5J 4 is a calcium release-activated calcium (CRAC) channel blocker. Blocks store-operated calcium entry (SOCE) in HeLa-O+S cells and Th17 cells in vitro. Ameliorates symptoms and reduces CNS infiltration by mononuclear cells in a mouse model of experimental autoimmune encephalomyelitis (EAE) in vivo.
Specifications
Specifications
| Chemical Name or Material | N-[[(6-Hydroxy-1-naphthalenyl)amino]thioxomethyl]-2-furancarboxamide |
| CAS | 827001-82-1 |
| Quantity | 10 mg |
| Target | Other Calcium Channel Blockers |
| Molecular Formula | C16H12N2O3S |
| Formula Weight | 312.34 |
| Purity | >98% |