missing translation for 'onlineSavingsMsg'
Learn More

Huber Kaeltmaschinenbau™ 5-way Header With Ball Valve

5-Way header with ball valves M16 x 1

Brand:  Huber Kaeltmaschinenbau™ 6816

Additional Details : Weight : 0.52000kg

Product Code. 10692563

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

  • 5-way header with ball valves, M16 x 1
  • Temperature range: -20°C to +140°C
  • Max. 6bar at +140°C
Specifications

Specifications

-20°C to +140°C
5-way Header with Ball Valves
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers