Learn More
IBA Solutions For Life Science 5-Methyl-2'-dCTP
A modified nucleoside-triphosphate (3'-modified oligos) that allows researchers to study the inhibition of DNA polymerases and DNA-protein interactions.
Brand:  IBA Solutions For Life Science 5-0612-265
Additional Details : Weight : 0.00050kg
 View more versions of this product
Product Code. 15623638
  • 819.00€ /
Estimated Shipment date
from Supplier 28-05-2019
Add to basket
Description
Description
IBA has a nucleoside-triphosphate synthesis facility that offers an extensive range of triphosphates for many applications.
  • All products are double HPLC purified, desalted and well characterized by HPLC and 31P-NMR
  • They are also available as monophosphates and nucleosides
Specifications
Specifications
Yes
To study the inhibition of DNA polymerases and DNA-protein interactions
5.0 μmol
Yes
Nucleoside-triphosphate
Videos