missing translation for 'onlineSavingsMsg'
Learn More

5-Methoxy-2-mercaptobenzimidazole, 99+%, Thermo Scientific Chemicals

Catalog Number p-157772
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
37052-78-1
C8H8N2OS
180.23
MFCD00134581
KOFBRZWVWJCLGM-UHFFFAOYSA-N
5-methoxy-2-mercaptobenzimidazole, 2-mercapto-5-methoxybenzimidazole, 5-methoxy-1h-benzo d imidazole-2-thiol, 5-methoxy-2-benzimidazolethiol, 5-methoxy-1h-benzimidazole-2-thiol, 5-methoxy-1h-1,3-benzodiazole-2-thiol, 2-mercapto-5-methoxy-1h-benzimidazole, 5-methoxy-1h-benzoimidazole-2-thiol, 1,3-dihydro-5-methoxy-2h-benzimidazole-2-thione, 5-methoxybenzimidazole-2-thiol
665603
5-methoxy-1,3-dihydrobenzimidazole-2-thione
COC1=CC=C2NC(=S)NC2=C1
This item is not returnable. View return policy
37052-78-1
C8H8N2OS
180.23
MFCD00134581
KOFBRZWVWJCLGM-UHFFFAOYSA-N
5-methoxy-2-mercaptobenzimidazole, 2-mercapto-5-methoxybenzimidazole, 5-methoxy-1h-benzo d imidazole-2-thiol, 5-methoxy-2-benzimidazolethiol, 5-methoxy-1h-benzimidazole-2-thiol, 5-methoxy-1h-1,3-benzodiazole-2-thiol, 2-mercapto-5-methoxy-1h-benzimidazole, 5-methoxy-1h-benzoimidazole-2-thiol, 1,3-dihydro-5-methoxy-2h-benzimidazole-2-thione, 5-methoxybenzimidazole-2-thiol
665603
5-methoxy-1,3-dihydrobenzimidazole-2-thione
COC1=CC=C2NC(=S)NC2=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Absorbance (1% soln. in methanol) (1 cm cell) no other absorption observed from 360 to 600nm, (1% soln. in methanol) (1 cm cell) no other absorption observed from 360 to 600nm at 0nm, 0.4 max. at 360nm
CAS Min % 99.0
CAS Max % 100.0
Melting Point 255°C to 262°C
Color Brown-Yellow to White
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 99% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in sodium hydroxide solution
Formula Weight 180.23
Percent Purity 99+%
Physical Form Powder
Chemical Name or Material 5-Methoxy-2-mercaptobenzimidazole, 99+%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'