missing translation for 'onlineSavingsMsg'
Learn More

5-Iodovanillin, 97%, Thermo Scientific Chemicals

Catalog Number p-149599
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
5438-36-8
C8H7IO3
278.04
MFCD00006941
FBBCSYADXYILEH-UHFFFAOYSA-N
5-iodovanillin, 5 iodovanillin, benzaldehyde, 4-hydroxy-3-iodo-5-methoxy, 5-iodo vanillin, 3-methoxy-4-hydroxy-5-iodobenzaldehyde, 5-iodovanilline, 5-iodovanilin, 5-iodo vaniline, acmc-1ao68
79499
4-hydroxy-3-iodo-5-methoxybenzaldehyde
COC1=C(C(=CC(=C1)C=O)I)O
This item is not returnable. View return policy
5438-36-8
C8H7IO3
278.04
MFCD00006941
FBBCSYADXYILEH-UHFFFAOYSA-N
5-iodovanillin, 5 iodovanillin, benzaldehyde, 4-hydroxy-3-iodo-5-methoxy, 5-iodo vanillin, 3-methoxy-4-hydroxy-5-iodobenzaldehyde, 5-iodovanilline, 5-iodovanilin, 5-iodo vaniline, acmc-1ao68
79499
4-hydroxy-3-iodo-5-methoxybenzaldehyde
COC1=C(C(=CC(=C1)C=O)I)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 180°C to 184°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Linear Formula IC6H2(OCH3)(OH)CHO
Quantity 5 g
Beilstein 08, 260
Formula Weight 278.04
Percent Purity 97%
Physical Form Fine Crystalline Powder
Chemical Name or Material 5-Iodovanillin, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 5-Iodovanillin
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'