Learn More
5-chloro-3-(chloromethyl)-1,2,4-thiadiazole, 97%, Thermo Scientific™
Catalog Number p-2702719
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
74461-64-6
C3H2Cl2N2S
169.02
MFCD00096470
FFFMIFUBJZBTCG-UHFFFAOYSA-N
5-chloro-3-chloromethyl-1,2,4-thiadiazole, 5-chloro-3-chloromethyl-1,2,4 thiadiazole, 3-chloromethyl-5-chloro-1,2,4 thiadiazole, 3-chloromethyl-5-chloro-1,2,4-thiadiazole, 1,2,4-thiadiazole,5-chloro-3-chloromethyl, 1,2,4-thiadiazole, 5-chloro-3-chloromethyl-7ci, 9ci
332610
5-chloro-3-(chloromethyl)-1,2,4-thiadiazole
ClCC1=NSC(Cl)=N1
74461-64-6
C3H2Cl2N2S
169.02
MFCD00096470
FFFMIFUBJZBTCG-UHFFFAOYSA-N
5-chloro-3-chloromethyl-1,2,4-thiadiazole, 5-chloro-3-chloromethyl-1,2,4 thiadiazole, 3-chloromethyl-5-chloro-1,2,4 thiadiazole, 3-chloromethyl-5-chloro-1,2,4-thiadiazole, 1,2,4-thiadiazole,5-chloro-3-chloromethyl, 1,2,4-thiadiazole, 5-chloro-3-chloromethyl-7ci, 9ci
332610
5-chloro-3-(chloromethyl)-1,2,4-thiadiazole
ClCC1=NSC(Cl)=N1
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 169.03
Percent Purity 97%
Chemical Name or Material 5-chloro-3-(chloromethyl)-1, 2, 4-thiadiazole