missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Bromo-4-chloro-3-indolyl-beta-D-galactopyranoside (X-GAL), >99%, Ultrapure, Thermo Scientific™
Click to view available options
Quantity:
100 mg
250 mg
1 g
2 g
5 g
Packaging:
Plastic bottle
Description
A histochemical substrate for β-Galactosidase. A substrate for blue/white colony screening. Effective concentration is approximately 50 μg/ml.
Specifications
Melting Point | ∼236°C (decomposition) |
Packaging | Plastic bottle |
Quantity | 100 mg |
Beilstein | 1402009 |
Sensitivity | Light sensitive |
Formula Weight | 408.63 |
Percent Purity | ≥99% |
Grade | Ultrapure |
Physical Form | Crystalline Powder |
Chemical Name or Material | 5-Bromo-4-chloro-3-indolyl-beta-D-galactopyranoside (X-GAL) |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'