Learn More
5-Bromo-1H-benzimidazole, 97%, Thermo Scientific™
Catalog Number p-4510685
Click to view available options
Quantity:
1 g
5 g
10 g
Packaging:
Amber Glass Bottle
4887-88-1
C7H5BrN2
197.04
MFCD00160001
GEDVWGDBMPJNEV-UHFFFAOYSA-N
5-bromo-1h-benzimidazole, 5-bromo-1h-benzo d imidazole, 5-bromo-1h-1,3-benzodiazole, 5-bromobenzimidazole, 1h-benzimidazole, 5-bromo, 5-bromo-3h-1,3-benzodiazole, 6-bromobenzimidazole, 5-bromo-benzimidazole, 6-bromo benzimidazole, pubchem9618
785299
BrC1=CC=C2N=CNC2=C1
4887-88-1
C7H5BrN2
197.04
MFCD00160001
GEDVWGDBMPJNEV-UHFFFAOYSA-N
5-bromo-1h-benzimidazole, 5-bromo-1h-benzo d imidazole, 5-bromo-1h-1,3-benzodiazole, 5-bromobenzimidazole, 1h-benzimidazole, 5-bromo, 5-bromo-3h-1,3-benzodiazole, 6-bromobenzimidazole, 5-bromo-benzimidazole, 6-bromo benzimidazole, pubchem9618
785299
BrC1=CC=C2N=CNC2=C1
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
IUPAC Name 6-bromo-1H-1,3-benzodiazole
Formula Weight 197.03
Percent Purity 97%
Chemical Name or Material 5-Bromo-1H-benzimidazole